Learn More
GE Healthcare Hyclone™ HyClone™ Irradiated Super Low IgG Fetal Bovine Serum
Processed to substantially reduce IgG (via Protein G chromatography) level, HyClone super low IgG fetal bovine serum (FBS) is suitable for use in more exacting applications.
Brand: GE Healthcare Hyclone™ SH30898.03IR
This item is not returnable.
View return policy
Description
- FBS contains natural levels of immunoglobulin (IgG) that may be too high for certain cell culture and protein purification applications.
- Serum from individual animals may be screened for low content and pooled to produce large batches of FBS with lower levels of IgG. However, this may be quite a challenge due to the scarcity of such low IgG animals and the logistics involved with the screening and collection process.
- Even naturally occurring low levels of IgG may still be too high for certain applications.
- Comparative levels of IgG in bovine serum products are shown in the table below.
- HyClone® Super Low IgG FBS performance is equivalent to non-processed FBS and is suitable for applications requiring lower levels of specific antibody, such as propagation of bovine viruses and monoclonal antibody production.
Specifications
–10°C | |
United States | |
Fetal Bovine Serum | |
500 mL |
Product Content Correction
Product Title
GE Healthcare Hyclone™ HyClone™ Irradiated Super Low IgG Fetal Bovine Serum > Quantity: 500mL
Spot an opportunity for improvement?Share a Content Correction