missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Human Mesenchymal Stem Cell Screened
Augment human mesenchymal stem cell growth
Brand: Cytiva SH30070.03M
This item is not returnable.
View return policy
Description
- Selected from our highest Defined FBS
- Filtered through sequential 40nm (0.04μm) pore-size rated filters
- Endotoxin: ≤10 EU/mL; Hemoglobin ≤10mg/dL

Specifications
1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 40 nm | |
None | |
≤ 10 EU/mL | |
≤ 10 EU/mL | |
USDA Inspected |
EMEA Certified | |
Human Mesenchymal Stem Cell Screened Fetal Bovine Serum, US Origin | |
Human Mesenchymal Stem Cell Screened | |
Sterile | |
Defined, Specialty screened | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Human Mesenchymal Stem Cell Screened > 500mL
Spot an opportunity for improvement?Share a Content Correction