Learn More
Invitrogen™ OSMR Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human OSM-R. Oncostatin M is a pleiotropic cytokine that belongs to the IL-6 group of cytokines.
£174.00 - £392.00
Specifications
Accession Number | Q99650 |
---|---|
Assay Range | 312.5-20,000 pg/mL |
Assay Sensitivity | 312.5 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15510867
|
Invitrogen™
88-50330-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
15500867
|
Invitrogen™
88-50330-22 |
2 x 96 Tests |
£174.00
Pack of 2 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
The Human Oncostatin M Receptor (OSMR) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human OSMR. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human OSM-R Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
Q99650 | |
312.5 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
IL-31R-beta,IL-31RBB,PLCA1,OSMR | |
HRP | |
Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL | |
Oncostatin M receptor beta, IL-31 receptor subunit | |
1 hr. 45 min. |
312.5-20,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
9180 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
For Research Use Only