Learn More
Lonza™ Human Umbilical Vein Endothelial Cells (HUVEC), Pooled in EGM™−2
Pooled donor cells are ideal for reducing donor variability. Lonza™ Human Umbilical Vein Endothelial Cells (HUVEC), Pooled in EGM™−2 are isolated in EGM™−2 Media with growth factors.
Brand:  Lonza™ C2519A
Product Code. 11525859
  • £610.00 / Each
Explore more special offers
Description
Description
  • Cryopreserved HUVECs shipped as frozen primaries
  • Guaranteed through 15 population doublings
  • Express CD31/105 and von Williebrand Factor VIII
  • Positive for acetylated low density lipoprotein uptake
  • All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots
  • A Certificate of Analysis is provided for each cell lot purchased
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lonza™ Human Umbilical Vein Endothelial Cells (HUVEC), Pooled in EGM™−2 > Media Type: EGM-2 BulletKit Medium