missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD11B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550682
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total HSD11B2 protein.
There are at least two isozymes of the corticosteroid 11-beta-dehydrogenase, a microsomal enzyme complex responsible for the interconversion of cortisol and cortisone. The type I isozyme has both 11-beta-dehydrogenase (cortisol to cortisone) and 11-oxoreductase (cortisone to cortisol) activities. The type II isozyme, encoded by this gene, has only 11-beta-dehydrogenase activity. In aldosterone-selective epithelial tissues such as the kidney, the type II isozyme catalyzes the glucocorticoid cortisol to the inactive metabolite cortisone, thus preventing illicit activation of the mineralocorticoid receptor. In tissues that do not express the mineralocorticoid receptor, such as the placenta and testis, it protects cells from the growth-inhibiting and/or pro-apoptotic effects of cortisol, particularly during embryonic development. Mutations in this gene cause the syndrome of apparent mineralocorticoid excess and hypertension.Specifications
HSD11B2 | |
Polyclonal | |
Unconjugated | |
HSD11B2 | |
11(beta)-HSD2; 11-beta-HSD; 11-beta-HSD type II; 11-beta-HSD2; 11-beta-hydroxysteroid dehydrogenase type 2; 11-beta-hydroxysteroid dehydrogenase type II; 11-DH2; 11-HSD type II; 11HSD2; AME; AME1; corticosteroid 11-beta-dehydrogenase isozyme 2; -HSD11 type II; HSD11B2; HSD11K; HSD2; hydroxysteroid (11-beta) dehydrogenase 2; hydroxysteroid 11-beta dehydrogenase 2; Hydroxysteroid dehydrogenase, 11 beta type 2; NAD-dependent 11-beta-hydroxysteroid dehydrogenase; SDR9C3; Short chain dehydrogenase/reductase family 9C member 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3291 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P80365 | |
HSD11B2 | |
Synthetic peptide corresponding to a region derived from internal residues of human hydroxysteroid (11-beta) dehydrogenase 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |