Learn More
HRC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552120
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SHEEDDNDDD DVSTEYGHQA HRHQDHRKEE VEAVSGEHHH HVPDHRHQGH RDEEEDEDVS TERWHQGPQH VHHGLVDEEE EEEEITVQFG HYVASHQPRG HKSDEEDFQD EYKTEVPHHH HHRVPREEDE EVSAELG Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 51%.
May play a role in the regulation of calcium sequestration or release in the SR of skeletal and cardiac muscle.Specifications
HRC | |
Polyclonal | |
Unconjugated | |
HRC | |
HCP; histidine rich calcium binding protein; HRC; Sarcoplasmic reticulum histidine-rich calcium-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3270 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23327 | |
HRC | |
Recombinant protein corresponding to Human HRC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only