missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPRT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513692
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with non-human primate based on sequence homology.
The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.Specifications
HPRT1 | |
Polyclonal | |
Unconjugated | |
HPRT1 | |
C81579; HGPRT; HGPRTase; HPGRT; Hprt; HPRT 1; HPRT B; Hprt1; hprt1 protein; hypoxanthine guanine phosphoribosyl transferase; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; hypoxanthine phosphoribosyltransferase 1; hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; id:ibd1344; id:ibd5108; testicular tissue protein Li 89; wu:fc10g09; zgc:56221; zgc:86608 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
15452, 3251 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P00492, P00493 | |
Hprt, HPRT1 | |
KLH conjugated synthetic peptide between 150-178 amino acids from the C-terminal region of human HPRT1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |