missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565483
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: QTFPSLQPGAS TLQRPRSQKR AED Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.
Hoxb-1 maintains this function by acting very early during hindbrain neurogenesis to specify effectors of the sonic hedgehog and Mash1 signaling pathways. Hoxb2 is a homeodomain protein important in neural development that is also expressed during erythropoiesis, hindbrain development and normal human adult lung development. Hoxb2 may modulate the amount of gamma-globin mRNA expressed during development and differentiation. In addition, Hoxb2 plays an important role in the patterning of hindbrain and pharyngeal arches in the zebrafish.Specifications
HOXB2 | |
Polyclonal | |
Unconjugated | |
HOXB2 | |
AI894218; homeo box 2H; homeo box B2; homeo box B2-like; homeobox B2; homeobox protein Hox-2.8; Homeobox protein Hox-2H; homeobox protein Hox-B2; HOX2; Hox-2.8; HOX2H; Hoxb2; Hoxb2-like epididymal protein; Hoxbes2; K8; K8 home protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P14652 | |
HOXB2 | |
Recombinant Human HOXB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only