missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA6 Monoclonal Antibody (3A6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519105
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: GYDALRPFPA SYGASSLPDK TYTSPCFYQQ SNSVLACNRA SYEYGASCFY SDKDLSGASP SGSGKQRGPG DYLHFSPEQQ YKPDSSSGQG KALHDEGADR
HOXA6 Monoclonal antibody specifically detects HOXA6 in Human samples. It is validated for ELISA, Western Blot.Specifications
HOXA6 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P31267 | |
HOXA6 | |
HOXA6 (NP_076919, 31 a.a. ∽ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
3A6 | |
Unconjugated | |
HOXA6 | |
homeo box 1B; homeo box A6; homeobox A6; homeobox protein Hox-1.2; Homeobox protein Hox-1B; homeobox protein HOXA6; homeobox protein Hox-A6; homeobox protein M5-4; HOX1; HOX1.2; Hox-1.2; HOX1B; Hoxa6; Hoxa-6 | |
Mouse | |
Affinity chromatography | |
RUO | |
3203 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |