missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540459
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AAAAANQCRN LMAHPAPLAP GAASAYSSAP GEAPPSAAAA AAAAAAAAAA Sequence homology: Dog: 83%; Human: 100%; Mouse: 92%
HOXA13 Polyclonal antibody specifically detects HOXA13 in Human samples. It is validated for Western Blot.Specifications
HOXA13 | |
Polyclonal | |
Unconjugated | |
Hoxa13 | |
Hd; homeo box 1J; homeo box A13; homeobox A13; homeobox protein Hox-1.10; Homeobox protein Hox-1J; homeobox protein HOXA13; homeobox protein Hox-A13; HOX1; Hox-1.10; HOX1J; HOXA13; hypodactyly; RGD1562483; transcription factor HOXA13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3209 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P31271 | |
Hoxa13 | |
Synthetic peptide directed towards the N-terminal of human HOXA13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |