missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Hostess™ Tela Fold Hand Towels

Self-presenting, dispensed open and ready to use. Kimberly-Clark™ Hostess™ Telafold Hand Towels include a break every seven sheets to stop streaming, helping to reduce costs and waste.

Specifications

Product Type Folded Hand Towel
Length (Metric) 24 cm
Certifications/Compliance EU Ecolabel
Format Tela Fold
For Use With (Application) Ideal for busy washrooms, for example in schools and other public facilities where economy is important
View More Specs

Description

Description

  • Small, 1-ply hand towels
  • Self-presenting, dispensed open and ready to use
  • Break every seven sheets stops streaming and helps to reduce costs and waste
  • Ideal for busy washrooms -- in schools and other public facilities where economy is important.
  • Carries the EU Ecolabel
Specifications

Specifications

Folded Hand Towel
EU Ecolabel
Ideal for busy washrooms, for example in schools and other public facilities where economy is important
Small
24 cm
Tela Fold
1-ply
Videos
SDS
Documents

Documents

Certificates