Learn More
Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors
For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.
POA
Specifications
Certifications/Compliance | ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC |
---|---|
Depth (English) | 1.1 in. |
Depth (Metric) | 2.8 cm |
Height (English) | 3.75 in. |
Height (Metric) | 9.52 cm |
Description
For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Secure and accurate history with datalogging with passcode protection
- Continuous LCD shows real-time gas concentrations
- Simple automatic calibration procedure
- Equipped with internal vibrating alarm for high noise areas
- Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
- Replaceable 3V battery
- Water-resistant
- Bright wide-angled visual alarm bars
- Two-year field-replaceable battery (1.5 year life) and sensor
- Adjustable alarm setpoints
- Multilanguage display options
- Compatible with Sampler motorized pump
- Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
- Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
- Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
- Weight: 82g (2.9 oz.)
Applications:
For single-gas detection of H2S, SO2, HCN, CO, NH3, Cl2, NO, NO2, PH3, ETO, ClO2, O3, O2. (See Product Type at right.)
Warranty
Two years including sensor, except Ammonia (one year)
Specifications
ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC | |
2.8 cm | |
9.52 cm | |
2.9 oz. | |
2 in. |
1.1 in. | |
3.75 in. | |
2 Years | |
83g | |
5.08 cm |