Learn More
HOMER3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559383
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ALVQTKDQEI QTLKSQTGGP REALEAAERE ETQQKVQDLE TRNAELEHQL RAMERSLEEA RAERERARAE VGRAAQLLDV RLFELSELRE GL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
This gene encodes a member of the HOMER family of postsynaptic density scaffolding proteins that share a similar domain structure consisting of an N-terminal Enabled/vasodilator-stimulated phosphoprotein homology 1 domain which mediates protein-protein interactions, and a carboxy-terminal coiled-coil domain and two leucine zipper motifs that are involved in self-oligomerization. The encoded protein binds numerous other proteins including group I metabotropic glutamate receptors, inositol 1,4,5-trisphosphate receptors and amyloid precursor proteins and has been implicated in diverse biological functions such as neuronal signaling, T-cell activation and trafficking of amyloid beta peptides. Alternative splicing results in multiple transcript variants.Specifications
HOMER3 | |
Polyclonal | |
Unconjugated | |
HOMER3 | |
AW146114; homer homolog 3; homer homolog 3 (Drosophila); homer protein homolog 3; homer scaffolding protein 3; Homer, neuronal immediate early gene, 3; HOMER3; homer-3; VASP/Ena-related gene up-regulated during seizure and LTP 3; Vesl3; Vesl-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9454 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NSC5 | |
HOMER3 | |
Recombinant protein corresponding to Human HOMER3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only