Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand:  Hoefer™ SE211A-5-.75
 View more versions of this product
Product Code. 12004797
  • £48.30 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
  • Lightweight
  • Virtually unbreakable
Specifications
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
PTFE
0.02 in.
9.8μL/ mm
0.51 in.
Comb
5
0.07 cm
1.3 cm
1.3 cm