missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15018-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs and also contains a nuclear localization motif. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
HNRNPR | |
Polyclonal | |
Unconjugated | |
HNRNPR | |
hnRNP R, HNRNPR, HNRPR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10236, 319110, 74326 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43390 | |
Hnrnpr | |
HNRNPR Fusion Protein Ag7028 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |