Learn More
HMGCS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529488
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Mouse liver. Predicted reactivity: Mouse (97%), Rat (97%), Pig (95%), Chicken (82%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.Specifications
HMGCS1 | |
Polyclonal | |
Unconjugated | |
HMGCS1 | |
3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 2; B130032C06Rik; HMG-CoA synthase; HMGCS; Hmgcs1; HMGCS1 protein-like; hydroxymethylglutaryl-CoA synthase 1; hydroxymethylglutaryl-CoA synthase, cytoplasmic; Unknown (protein for IMAGE:7955111) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
208715, 3157 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q01581, Q8JZK9 | |
HMGCS1 | |
Recombinant fragment corresponding to a region within amino acids 194 and 455 of Human HMGCS1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |