missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523362
Code : LD
Additional Details : Weight : 0.01000kg
Description
HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2, HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.Specifications
HMGB3 | |
Polyclonal | |
Unconjugated | |
HMGB3 | |
high mobility group box 3; high mobility group protein 1; high mobility group protein 2a; High mobility group protein 4; high mobility group protein B1; high mobility group protein B3; high mobilty group protein 2A; high-mobility group (nonhistone chromosomal) protein 4; high-mobility group protein 4; HMG-1; HMG2a; HMG-2a; HMG4; HMG-4; HMGB1; Hmgb3; HMGB3 transcript variant 1/2; MGC90319; RGD1564407; Unknown (protein for MGC:133786) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100069284, 15354, 3149, 396232, 481073, 613729 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
O15347, O54879, P40618, Q32L31 | |
HMGB3 | |
A portion of amino acids 150-200 of human HMGB3 | |
100 μg | |
Primary | |
Bovine, Canine, Chicken, Equine, Human, Mouse, Non-human Primate | |
Antibody | |
IgG |