Learn More
HLA-DR Monoclonal Antibody (TU36), Pacific Orange™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHLDR30
Code : 50
Additional Details : Weight : 0.50000kg
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DR, HLA-DR (Class II) | |
Monoclonal | |
Pacific Orange | |
HLA-DRB4 | |
AI323765; BLA-DRB3; BOLA-DRA; BoLA-DR-alpha; BoLA-DRB; BOLA-DRB3; BoLA-DRB3 protein; BoLA-DRB3.2; Bota-DRB01; Bota-DRB02; Bota-DRB04; Bota-DRB07; Bota-DRB21; Bota-DRB22; bovine leukocyte antigen; Cd74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; Cd74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; CLIP; Clone P2-beta-3; DASS-397D15.1; DHLAG; dinucleotide microsatellite; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DR beta; DLA-DRB1; DLA-DRBB1; DR beta-5; DR beta-chain antigen binding domain; DR-16; DR2-beta-2; DR4; DR-4; DR7; DR9; DR-9; DRB; DRB1; DRB1 transplantation antigen; DRB3; DRB4; DRBB1; DR-beta chain; DR-beta chain MHC class II; DRw10; Dw2; DW2.2/DR2.2; E-alpha-f; FLJ51114; FLJ75017; FLJ76359; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; H-2 cla | |
Mouse | |
purified | |
ASR | |
3122, 3123, 3125, 3126, 3127, 972 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
TU36 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P01903, P01911, P04233, P13762, P79483, Q30154 | |
CD74, HLA-DRA, HLA-DRB1, HLA-DRB3, HLA-DRB4, HLA-DRB5 | |
Human HLA-DR | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |