missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA class I ABC Mouse anti-Human, Porcine, Clone: 5C5B7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | HLA class I ABC |
---|---|
Clone | 5C5B7 |
Concentration | 1.11 mg/mL |
Applications | Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897505
|
Proteintech
66013-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887505
|
Proteintech
66013-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determits important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.Specifications
HLA class I ABC | |
1.11 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HLA A, HLA class I, HLA class I (HLA-A), HLA class I ABC, HLA-A, MHC class I antigen A*3 | |
HLA-A, LOC100515902 | |
IgG2b | |
Protein A | |
Antibody |
5C5B7 | |
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Porcine, Human | |
P30443 | |
100515902, 3105 | |
HLA class I ABC Fusion Protein Ag7488 | |
Primary | |
-20°C | |
HLA-A |