missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-ABC Mouse anti-Human, APC, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
£208.00 - £610.00
Specifications
Antigen | HLA-ABC |
---|---|
Clone | G46-2.6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15390468
|
BD Biosciences
562006 |
25 Tests |
£208.00
Each |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15808268
|
BD Biosciences
555555 |
100 Tests |
£610.00
Each |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The G46-2.6 monoclonal antibody specifically binds to a monomorphic epitope on the alpha chain of major histocompatibility (MHC) HLA Class I antigens. Class I MHC antigens are widely distributed on human nucleated cells.
Specifications
HLA-ABC | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
G46-2.6 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.