Learn More
Histamine diphosphate, 98%, Thermo Scientific Chemicals
Endogenous H1 and H2 agonist
£41.61 - £152.76
Chemical Identifiers
CAS | 51-74-1 |
---|---|
Molecular Formula | C5H15N3O8P2 |
Molecular Weight (g/mol) | 307.14 |
MDL Number | MFCD00064316 |
InChI Key | ZHIBQGJKHVBLJJ-UHFFFAOYSA-N |
Synonym | histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate |
PubChem CID | 65513 |
ChEBI | CHEBI:51193 |
SMILES | OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15493739
|
Thermo Scientific Alfa Aesar
J61088.06 |
5 g |
£41.61
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403749
|
Thermo Scientific Alfa Aesar
J61088.14 |
25 g |
£152.76
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHistamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Chemical Identifiers
51-74-1 | |
307.14 | |
ZHIBQGJKHVBLJJ-UHFFFAOYSA-N | |
65513 | |
OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1 |
C5H15N3O8P2 | |
MFCD00064316 | |
histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate | |
CHEBI:51193 |
Specifications
51-74-1 | |
White | |
MFCD00064316 | |
14,4719 | |
Soluble in water. | |
OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1 | |
307.14 | |
CHEBI:51193 | |
≥99% | |
Histamine diphosphate |
128°C to 132°C | |
C5H15N3O8P2 | |
Hygroscopic | |
histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate | |
ZHIBQGJKHVBLJJ-UHFFFAOYSA-N | |
2-(1H-imidazol-5-yl)ethanamine;phosphoric acid | |
65513 | |
307.14 | |
Powder |
Safety and Handling
GHS H Statement
H334-H318-H302-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H318-H334-H335
missing translation for 'einecsNumber' : 200-118-4
missing translation for 'rtecsNumber' : NI5425000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only