missing translation for 'onlineSavingsMsg'
Learn More

Hexane, mixture of isomers, Puriss., ≥85% isomers basis (GC), Honeywell™

Product Code. 15681490
Click to view available options
Quantity:
1 L
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Puriss.,≥85% isomers basis (GC)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.672g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.075
Flash Point −23°C (−9.4°F)
Ignition Point 437°F
UN Number 1208
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥85% isomers basis (GC)
Grade puriss.
Chemical Name or Material Hexane, Mixture Of Isomers
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'