Learn More
Heptanophenone, 98+%, Thermo Scientific Chemicals
£28.30 - £88.20
Chemical Identifiers
CAS | 1671-75-6 |
---|---|
Molecular Formula | C13H18O |
Molecular Weight (g/mol) | 190.29 |
MDL Number | MFCD00009539 |
InChI Key | UXMQORVHJMUQFD-UHFFFAOYSA-N |
Synonym | heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone # |
PubChem CID | 74282 |
IUPAC Name | 1-phenylheptan-1-one |
SMILES | CCCCCCC(=O)C1=CC=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1671-75-6 | |
190.29 | |
UXMQORVHJMUQFD-UHFFFAOYSA-N | |
74282 | |
CCCCCCC(=O)C1=CC=CC=C1 |
C13H18O | |
MFCD00009539 | |
heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone # | |
1-phenylheptan-1-one |
Specifications
1671-75-6 | |
0.946 | |
>110°C (230°F) | |
1.5077 | |
1865692 | |
UXMQORVHJMUQFD-UHFFFAOYSA-N | |
1-phenylheptan-1-one | |
74282 | |
≥98% |
∼17°C | |
283°C | |
C13H18O | |
MFCD00009539 | |
heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone # | |
CCCCCCC(=O)C1=CC=CC=C1 | |
190.29 | |
190.29 | |
Heptanophenone |
Safety and Handling
missing translation for 'einecsNumber' : 216-802-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.