Learn More
Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals
£34.87 - £277.38
Chemical Identifiers
CAS | 336-59-4 |
---|---|
Molecular Formula | C8F14O3 |
Molecular Weight (g/mol) | 410.06 |
MDL Number | MFCD00000432 |
InChI Key | UFFSXJKVKBQEHC-UHFFFAOYSA-N |
Synonym | heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization |
PubChem CID | 67643 |
ChEBI | CHEBI:39424 |
IUPAC Name | 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate |
SMILES | FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10144090
|
Thermo Scientific Acros
169260050 |
5 g | Glass bottle |
£34.87
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522961
|
Thermo Scientific Acros
169260250 |
25 g | Glass bottle |
£89.75
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10309143
|
Thermo Scientific Acros
169261000 |
100 g | Glass bottle |
£277.38
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagentChemical Identifiers
336-59-4 | |
410.06 | |
UFFSXJKVKBQEHC-UHFFFAOYSA-N | |
67643 | |
2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate |
C8F14O3 | |
MFCD00000432 | |
heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization | |
CHEBI:39424 | |
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F |
Specifications
336-59-4 | |
1.6500g/mL | |
Authentic | |
C8F14O3 | |
MFCD00000432 | |
heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization | |
UFFSXJKVKBQEHC-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate | |
67643 | |
410.05 | |
Liquid |
-43°C | |
108°C to 111°C | |
98+% | |
(CF3CF2CF2CO)2O | |
1.65 | |
Solubility in water: reacts with water forming heptafluorobutyric acid | |
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F | |
410.06 | |
CHEBI:39424 | |
98+% | |
Heptafluorobutyric anhydride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-410-8
- Heptafluorobutyric anhydride
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.