missing translation for 'onlineSavingsMsg'
Learn More

HEPES, Free Acid, Ultrapure Bioreagent, J.T. Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

£225.00 - £810.00

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15508214
View Documents
Avantor J.T.Baker
4018-01
100 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15518214
View Documents
Avantor J.T.Baker
4018-04
500 g Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

    For Liquid Chromatography and Molecular Biology applications; For Laboratory,Research,or Manufacturing Use
Specifications

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
CHEBI:42334
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid

Specifications

7365-45-9
MFCD00006158
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
Ultrapure Bioreagent
C8H18N2O4S
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
238.31
HEPES, Free Acid, Free Acid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.