missing translation for 'onlineSavingsMsg'
Learn More
Heparin Sodium Salt, Thermo Scientific Chemicals Product Code.: 15885548

Heparin Sodium Salt, Thermo Scientific Chemicals

Product Code. 15885548 Shop All Thermo Scientific Temperature Control Solutions Products
100 kU, Plastic bottle
Click to view available options
Quantity:
1 mU
100 kU
500 kU
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15885548

Brand: Thermo Scientific J16920.!P

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 9041-08-1

Specifications

Physical Form Powder
Quantity 100 kU
MDL Number MFCD00081689
Packaging Plastic bottle
Chemical Name or Material Heparin Sodium Salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heparin Sodium Salt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.