Learn More
BINDER™ HEPA Air Filter
BINDER™ HEPA Air Filter is an accessory for unit FED 400 Drying and Heating Chamber.
Brand:  BINDER™ 8012-0076
1061.15 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 1.00000kg
Product Code. 15691666
  • £1420.00 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 μm)
Order Info
ForUseWithApplication
Specifications
Specifications
HEPA air filter - On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% (at 0.3 μm))
BINDER Chambers FED 400
HEPA Filter