missing translation for 'onlineSavingsMsg'
Learn More

HEMGN Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA554037

Product Code. 15751971

  • £418.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: GKDQSHLKHH QTPDPHQEEN HSPEVIGTWS LRNRELLRKR KAEVHEKETS QWLFGEQKKR KQQRTGKGNR RGRKRQQNTE LKVEPQPQIE KEIVEKALAP I Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 62%.

HEMGN regulates the proliferation and differentiation of hematopoietic cells. Overexpression of HEMGN block the TPA-induced megakaryocytic differentiation in the K562 cell model. HEMGN may also prevent cell apoptosis through the activation of the nuclear factor-kappa B (NF-kB).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HEMGN
Polyclonal
Unconjugated
HEMGN
4921524M03Rik; AI317176; CT155; EDAG; EDAG-1; erythroid differentiation-associated gene protein; Hemgn; Hemogen; hemopoietic gene protein; Hgn; mNDR; NDR; negative differentiation regulator protein; nuclear protein; PRO1037; PRO1620; Unknown (protein for MGC:134079)
Rabbit
Antigen affinity chromatography
RUO
55363
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.7 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BXL5
HEMGN
Recombinant protein corresponding to Human HEMGN
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only