Learn More
Brady™ PermaSleeve™ Heat-shrink Polyolefin Wiremarker Sleeves
Bulk roll of PermaSleeve markers. PermaSleeve™ Heat-shrink Polyolefin Wiremarker Sleeves provide a wire marking solution that combines speed, ease of use, and permanence to create long-lasting, high quality identification.
Brand:  Brady™ 115013
Code : NEW
Additional Details : Weight : 0.67130kg
 View more versions of this product
Product Code. 11791765
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • These Sleeves are self-extinguishing.
  • Meet the requirements of SAE AMS-DTL-23053/5 Class 1, SAE-AS-8153, MIL-STD-202 method 215K and is UL224 recognised.
  • CSA acceptance pending.
Specifications
Specifications
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL Recognised
Yellow
Roll
Single-side Printable
Wire and Cable Markers
BMP61, BMP71
-55°C
1000 Pk.
3:1
-55°C to +135°C
20 AWG - 10 AWG
1 in.
1 in.
8.51 mm
1.57 to 3.81 mm
Bulk PermaSleeve Wire Marker Sleeves for BMP61 and BMP71
Matte
Polyolefin
Thermal Transfer
Electrical labelling, Wire and cable labelling
1
1000 Sleeves/Box
Rectangle
-67°F to +275°F
19.05 mm
42.16 x 25.78 mm
25.4 mm
0.34 in.
0.06 to 0.15 in.