missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Health and safety law poster, vinyl, 594mm x 420mm
Brand: Manutan A352631
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Employers have a legal duty to display the HSE health and safety law poster in a prominent place in every workplace. Inform employees of their rights and obligations under Health and Safety Law. Space to add details of employee safety representatives.- Vinyl
- Dimensions (H x W) : 594mm x 420mm