missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Deep Ledge Utility Cart

Use these durable utility carts for a variety of general laboratory storage and transportation. Cole-Parmer™ Deep Ledge Utility Cart fabricated from a tough, high-density polyethylene and provide an excellent chemical resistance to most substances, including most acids and bases

Brand:  Cole-Parmer™ BC20303DMB

Product Code. 11775629

  • £476.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Very durable, made from enamelled steel deck and angle subframe with tough HDPE shelves
  • 2 and 3 shelf carts manufactured with Microban™, an antimicrobial product that protects shelves from bacteria, mould, and mildew
  • Smooth, wipe clean shelves feature a deep, 44.5mm lip to contain spills
  • Centre shelves of 3 and 4 shelf models adjust at 25.4mm increments
  • With swivel castors
  • Handle extends 711mm above deck
  • Capacity 181kg
Specifications

Specifications

Trolley
831.85 mm
1
1041.4 mm
181 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers