missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ HDPE, Carboy
Closure Material: HDPE
Brand: Cole-Parmer™ 06032-20
This item is not returnable.
View return policy
Specifications
Not Autoclavable | |
HDPE | |
Needle-type spigot | |
1 Ea. | |
5-3/4 in. |
9 L | |
13.5 in. | |
HDPE | |
Carboy |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction