missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Hazardous storage cabinet, for acid/alkali, white, 915mm x 915mm x 459mm

Brand:  Key Industrial Equipment™ A350597

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12881527

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Hard wearing cabinets suitable for storing mild acids and corrosive chemicals.
  • One shelf
  • Dimensions (H x W x D) : 915mm x 915mm x 459mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers