Learn More
BTX™ Mini Micro-Grabber Cables for Electroporators, Positive and Negative
Use these Mini Micro-Grabber Cables to connect external devices (Genetrodes, Genepaddles, Microslides, Meander Fusion Chamber) to BTX electroporation generators.
Brand: BTX™ 450503
This item is not returnable.
View return policy
Description
- Multiple purposes based on electrode type of interest
- Compatible with most BTX Systems such as ECM 2001+ and Gemini X2
Specifications
Tissue Slice Chamber, Oocyte, L-Shaped Genetrodes Electrodes | |
100 V | |
Non-sterile | |
Digital User |
Adapter cable | |
Mini Micro-Grabber Cables, Positive and Negative | |
Banana Plug to Micrograbber | |
Square Wave |
Product Content Correction
Product Title
BTX™ Mini Micro-Grabber Cables for Electroporators, Positive and Negative > Mini Micro-Grabber Cables; Type: Adapter Cable
Spot an opportunity for improvement?Share a Content Correction