Learn More
IKA™ H 99 Protective Cover
Silicone in-use protective cover for stirrers is resistant to most acids, alkaline solutions, and organic solvents.
Brand:  IKA™ 0002734500
Product Code. 10356971
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For RET series and RCT Basic magnetic stirrers
  • Form-fitting, in-use cover
  • Resistant to most acids, alkaline solutions, and organic solvents
  • Silicone construction
Specifications
Specifications
RET series and RCT Basic magnetic stirrer
H 99
Up to 135°C
In-use Cover