Learn More
Cytiva GV050 Lower Part
This Lower Part is a replacement part for Whatman™ GV 050 Series Vacuum Filtration Manifolds.
Brand:  Cytiva 10442212
Product Code. 11344944
  • £174.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
  • For Use With (Application): Vacuum Filtration
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Reusable
Borosilicate Glass
Vacuum Filtration
1 Ea.
Filter Funnel
Filter Funnel (Base Section, NS45)
Whatman GV050/2 Glass Vacuum Filter Holder
Non-sterile
GV050/2/12
GV050