missing translation for 'onlineSavingsMsg'
Learn More

Griffin Education Calorimeter block cylindrical 115mm x 38mm

Brand:  Griffin Education 6127

Calorimeter block cylindrical 115mm x 38mm

Code : 07

Additional Details : Weight : 1.03400kg

Product Code. 13106330

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers