Learn More
Ansell™ Comfort G26G Nitrile Immersion Gloves
Excellent resistance to many solvents and oil based chemicals. Flexible and comfortable for an excellent dexterity. Superior raised grip pattern for safety handling.
Brand:  Ansell™ A340085
 View more versions of this product
Product Code. 13497606
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Very good abrasion resistance.
  • Allows contact up to 100°C.
  • Superior raised grip pattern for safety handlings.
  • Open cuff with textile edge binding allows ventilation to keep hand cool.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrile
Immersion Gloves
Green
Cotton
12 Pairs
Nitrile
8.5
Gauntlet
COMFORT G26G
Excellent resistance to many solvents and oil based chemicals