missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPKOW Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14728-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.Specifications
GPKOW | |
Polyclonal | |
Unconjugated | |
GPKOW | |
G patch domain and KOW motifs, GPATC5, GPATCH5, GPKOW, Protein MOS2 homolog, Protein T54, T54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209416, 27238, 679890 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q56A08, Q92917 | |
GPKOW | |
GPKOW Fusion Protein Ag6487 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |