missing translation for 'onlineSavingsMsg'
Learn More

Gold, plasma standard solution, Specpure™ Au 10,000μg/mL, Thermo Scientific Chemicals

£284.42 - £1433.13

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322318
View Documents
Thermo Scientific Alfa Aesar
014392.AD
50 mL
£284.42
50mL
Please to purchase this item. Need a web account? Register with us today!
11312318
View Documents
Thermo Scientific Alfa Aesar
014392.AE
100 mL
£424.11
100mL
Please to purchase this item. Need a web account? Register with us today!
11400430
View Documents
Thermo Scientific Alfa Aesar
014392.AP
500 mL
£1433.13
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gold, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold

Specifications

7440-57-5
MFCD00003436
[Au]
196.97
CHEBI:30050
Immiscible with water
Gold
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Characteristic
Liquid
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: II

missing translation for 'einecsNumber' : 231-595-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.