missing translation for 'onlineSavingsMsg'
Learn More

Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, positively charged, Thermo Scientific™

Product Code. 15435015
Click to view available options
Quantity:
5 mL
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Colorless
Boiling Point >60°C (decomposition)
Quantity 5 mL
Odor Odorless
Form Liquid
Chemical Name or Material Gold nanorods, 19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, positively charged
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'