Learn More
Goat anti-Rabbit IgG, Secondary Antibody, Abcam™
Brand: Abcam AB6717-1MG
Code : Z1
Additional Details : Weight : 0.00001kg
Description
Host Species: Goat
Isotype: IgG
Cross Reactivity: Rabbit
Immunogen: Rabbit IgG whole molecule
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy
Specifications
Rabbit IgG | |
2mg/mL | |
Goat | |
1 mg | |
Rabbit |
Polyclonal | |
FITC | |
Affinity Purified | |
Secondary | |
IgG |