missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG whole molecule, Biotin, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10748.2mg
This item is not returnable.
View return policy
Specifications
Goat anti-Mouse IgG whole molecule | |
Polyclonal | |
Goat anti-mouse IgG recognizes mouse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin. | |
In 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (Immunoglobulin and Protease free)) | |
Mouse IgG whole molecule | |
2 mg | |
Mouse | |
Lyophilized |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Biotin | |
The optimal working dilution should be determined by the end user. | |
Goat | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction