missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL3 Mouse anti-Human, Unconjugated, Clone: 3B8F10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | GNL3 |
---|---|
Clone | 3B8F10 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829655
|
Proteintech
67169-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819655
|
Proteintech
67169-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GNL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNL3. This antibody recognizes human, mouse, rat antigen. The GNL3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
GNL3 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
26354 | |
Primary |
3B8F10 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title