Learn More
Sampling Systems™ GMP Pallet Knife
These stainless steel GMP pallet knives are ideal for a wide range of applications, including cleaning, scraping and mixing. Made from medical grade 420 stainless steel, they are crevice-free and have a polished finish for easy cleaning after use.
Brand:  Sampling Systems™ A123-350
 View more versions of this product
Product Code. 17942092
  • £170.05 / Each
Explore more special offers
Description
Description
  • Strong and flexible
  • Range of different lengths
Specifications
Specifications
Non-sterile
Curved
Pallet Knife
490 mm
Autoclavable
Reusable
Stainless Steel