missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid monohydrate, 98%, pure, Thermo Scientific Chemicals

£26.23 - £212.06

Chemical Identifiers

CAS 563-96-2
Molecular Formula C2H2O3·H2O
Molecular Weight (g/mol) 92.06
MDL Number MFCD00127974
InChI Key MOOYVEVEDVVKGD-UHFFFAOYSA-N
Synonym glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
PubChem CID 15620607
IUPAC Name oxaldehydic acid;hydrate
SMILES C(=O)C(=O)O.O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10182480
View Documents Promotion Details
Thermo Scientific Acros
120170250
25 g Plastic bottle
£26.23
25g
Please to purchase this item. Need a web account? Register with us today!
10647652
View Documents
Thermo Scientific Acros
120171000
100 g Plastic bottle
£60.70
100g
Please to purchase this item. Need a web account? Register with us today!
10500941
View Documents
Thermo Scientific Acros
120175000
500 g Plastic bottle
£212.06
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

563-96-2
92.06
MOOYVEVEDVVKGD-UHFFFAOYSA-N
15620607
C(=O)C(=O)O.O
C2H2O3·H2O
MFCD00127974
glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
oxaldehydic acid;hydrate

Specifications

563-96-2
White to Yellow
Authentic
C2H2O3·H2O
MFCD00127974
Solubility in water: freely soluble.
C(=O)C(=O)O.O
92.06
92.06
Pure
Glyoxylic acid monohydrate, 98%
48.0°C to 52.0°C
>110°C
98%
HCOCO2H·H2O
glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
MOOYVEVEDVVKGD-UHFFFAOYSA-N
oxaldehydic acid;hydrate
15620607
98%
Crystalline Powder and/or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti

GHS Signal Word: Danger

Product Identifier
  • Glyoxylic acid monohydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.