Learn More
GLRA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13831-1-AP-150UL
 View more versions of this product
Product Code. 16842224
  • £349.00 / 150µL
Explore more special offers
Description
Description
Glycine is an inhibitory neurotransmitter in the central nervous system. The glycine receptor (GlyR) is a member of the nicotinic acetylcholine receptor of ligand-gated ion channels. This receptor has important roles in a variety of physiological processes, especially in mediating inhibitory neurotransmission in the spinal cord and brain stem. GlyR is a pentameric receptor comprised of alpha and beta subunits. Four alpha-subunits (GLRA1, GLRA2, GLRA3, GLRA4) and one beta-subunit (GLRB) of GlyR have been identified.
Specifications
Specifications
GLRA2
Polyclonal
Unconjugated
GLRA2
GLR, GLRA2, glycine receptor, alpha 2
Rabbit
Antigen Affinity Chromatography
RUO
237213, 24397, 2742
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22771, P23416, Q7TNC8
GLRA2
GLRA2 Fusion Protein Ag4743
150 μL
Primary
Human, Mouse, Rat