Learn More
Gibco™ DPBS, calcium, magnesium
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 14040133
17.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red
Specifications
No Sodium Pyruvate | |
Storage conditions: 2-8° C Shelf life: 36 months from date of manufacture |
|
Gibco™ | |
500 mL | |
Washing Cells Before Dissociation, Transporting Cells or Tissue Samples, Diluting Cells for Counting, and Preparing Reagents | |
270 - 300 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Animal Origin-Free | |
1 X | |
DPBS | |
Dulbecco's Phosphate Buffered Saline | |
Sustainable packaging | |
7.0 to 7.2 | |
Gel Ice | |
Liquid |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.