missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bovine Serum, New Zealand origin
US-qualified performance sera for general cell culture with common cell lines
£29.21 - £82.50
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | 12 to 24 months |
Species | Cattle/Bovine |
Country of Origin | New Zealand |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11520526
|
Gibco™
16170086 |
100 mL |
£29.21
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11510526
|
Gibco™
16170078 |
500 mL |
£82.50
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sera Category: Secure
- Origin: New Zealand
- Collected from cattle of age 12-24 months
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Adult Bovine Serum | |
Sterile-filtered | |
Gibco™ |
12 to 24 months | |
New Zealand | |
Bottle | |
Sterile-filtered | |
Sustainable packaging | |
Frozen |