missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYTTD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24560-1-AP-20UL
This item is not returnable.
View return policy
Description
FYTD1 required for mRNA export from the nucleus to the cytoplasm. This protein acts as an adapter that uses the BAT1/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. FYTD1 associates with spliced and unspliced mRNAs simultaneously with THOC4.Specifications
FYTTD1 | |
Polyclonal | |
Unconjugated | |
FYTTD1 | |
FYTTD1, Protein 40 2 3, UAP56 interacting factor, UIF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84248 | |
-20°C | |
Liquid |
Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96QD9 | |
FYTTD1 | |
FYTTD1 Fusion Protein Ag20051 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |